Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Lady Beetles
SDPSCI_ED

Catalog No. S07642ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
100/Container
Catalog No. Quantity
S07642ND 100/Container
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S07642ND

Supplier: Carolina Biological Supply 144222

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Well-known predator of aphids.

  • Hippodamia convergens
  • Often used for biological pest control

Specifications

For Use With (Application) Well known predator of aphids, often used for biological pest control.
Storage Requirements 18°C
Quantity 100/Container
Scientific Name Hippodamia convergens
Sufficient For Class of 30 students
Product Type Lady Beetle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.