Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Live Brine Shrimp
SDPSCI_ED

Catalog No. S07598ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
2 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07598ND 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07598ND Supplier Carolina Biological Supply Supplier No. 142230

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

A typical primitive arthropod with a segmented body to which is attached broad leaf-like appendages.

  • Ideal food for all fish, aquatic insects, and amphibians, without the trouble of hatching
  • Two ounces
  • With instructions

Specifications

For Use With (Application) Ideal food for all fish, aquatic insects and amphibians, without the trouble of hatching.
Storage Requirements 26°C to 28°C
Quantity 2 oz.
Scientific Name Artemia
Includes Instructions
Sufficient For Class of 30 students
Product Type Brine Shrimp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.