Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Painted Lady Culture 33 Larvae Set
SDPSCI_ED

Catalog No. S07711CND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
3/Container
Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07711CND Ea.
S07710ND 3/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07711CND Supplier Carolina Biological Supply Supplier No. 144072

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • 1 cup of 33 larvae
  • 1 cup of food

Great for use as refills for butterfly kits!

This larvae set is perfect for those who enjoy butterflies but do not need a complete learning kit.

  • Quantity of food is only sufficient for 33 larvae
  • Larvae Set does not include the Culture Cup Set
  • Culture Cup Set must be purchased separately

Note: Additional larvae are supplied with each Larvae/Food Set to ensure that at least 33 larvae emerge as healthy adults. Extra food must be purchased if you wish to maintain additional larvae.

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

For Use With (Application) Great for use as refills for butterfly kits.
Quantity Ea.
Includes 1 cup of 33 larvae, 1 cup of food
Sufficient For Class of 30 students
Product Type Painted Lady Butterfly Culture 33 Larvae Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.