Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Painted Lady Culture Cup Set
SDPSCI_ED

Catalog No. S07711 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07711 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07711 Supplier Carolina Biological Supply Supplier No. 144068

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

For those who enjoy butterflies but do not need a complete learning kit.

Great for use as refills for butterfly kits!

  • Enough cups, lids, and culture tissue to accommodate one Painted Lady 33-Larvae Set

Specifications

For Use With (Application) Great for use as refills for butterfly kits.
Quantity 1 Ea.
Includes Enough cups, lids and culture tissue to accommodate one painted lady 33 larvae set
Sufficient For Class of 30 students
Product Type Painted Lady Butterfly Culture Cup Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.