Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Painted Lady Culture Medium
SDPSCI_ED

Catalog No. S07712ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
2/Container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07712ND 2/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07712ND Supplier Carolina Biological Supply Supplier No. 144040

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

A unique innovation, this culture medium provides all the ready-to-use nutrition your caterpillars require to grow.

There is no food to mix. Simply divide the medium into smaller containers, such as the Culture Cup Set (S07711). Caterpillars will eat the culture medium supplied in each culture vessel. One cup of medium feeds 33 larvae; two cups feed up to 75 larvae.

  • Two cups
  • Each cup contains 8 oz. of medium
  • Includes instructions

Note: Culture cups not included; refrigerate medium for up to two weeks.

Specifications

For Use With (Application) This culture medium provides all the ready-to-use nutrition your caterpillarss require to grow.
Storage Requirements 22°C to 25°C
Quantity 2/Container
Includes Two cups, each cup contains 8 oz. of medium, with instructions
Sufficient For Class of 30 students
Product Type Painted Lady Butterfly Culture Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.