Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Pond Mystery Mix
SDPSCI_ED

Catalog No. S07453ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07453ND Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07453ND Supplier Carolina Biological Supply Supplier No. 132010A

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • Bacteria
  • Protozoa
  • Algae
  • Rotifers
  • Ostracods
  • And others
  • Instructions and illustrations of some of the more commonly seen organisms

Investigate the diversity of pond microorganisms with this unique dried mixture; just add springwater to bring it to life!

The mixture contains a variety of materials and nutrients to support microorganism growth and food chains. Your students can observe bacteria and other single-celled organisms within 24 hours after adding water; larger microinvertebrates will appear within a week.

  • Safely stores for years in dried form without special handling

Specifications

Storage Requirements 20°C to 22°C
Quantity Ea.
Includes Bacteria, Protozoa, Algae, Rotifers, Ostracods and others instructions and illustrations of some of the more commonly seen organisms
Sufficient For Class of 30 students
Product Type Pond Mystery Mix
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.