Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Protozoa Survey Mixture
SDPSCI_ED

Catalog No. S07455ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07455ND Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07455ND Supplier Carolina Biological Supply Supplier No. 131940

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • A single culture containing six representative genera of protozoa
  • Dichotomous key to aid in their identification
  • Materials for a class of 30 students

The protozoan body consists of a single cell, but it is a mistake to think of these organisms as simple.

  • Shipped in a single 2 oz. jar

Specifications

Packaging Jar
Storage Requirements 20°C to 22°C
Quantity Ea.
Includes A single culture containing 6 representative Protozoa and a simple dichotomous key to aid in their identification
Sufficient For Class of 30 students
Product Type Protozoa Survey Mixture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.