Learn More
Carolina™ Sea Urchin Embryology Kit
SDPSCI_ED
Catalog No. S07586ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
Ea.
Catalog No. Quantity
S07586ND Ea.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07586ND Supplier Carolina Biological Supply Supplier No. 162505
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes
  • 14 fertile sea urchins (Eucidaris tribuloides or Lytechinus variegatus)
  • 30mL 0.5M potassium chloride solution
  • 20 gamete shedding cups
  • 20 plastic Petri dishes
  • 20 pipets
  • Hypodermic syringes with needles
  • One gallon of seawater
  • Instructions
Everything your class needs to witness sea urchins develop is available in this one convenient kit.
  • Urchins are ripe at the time of arrival
  • Use the day of arrival or immediately the next day to avoid reabsorption of the gametes
  • Not sexed but male-to-female ratio is usually about 1:1
  • Includes all the materials and instructions necessary for achieving viable gametes and for following development from early cleavage through pluteus
Specifications
Quantity Ea.
Includes 14 fertile sea urchins (Eucidaris tribuloides or Lytechinus variegatus), 30mL 0.5M potassium chloride solution, 20 gamete shedding cups, 20 plastic petri dishes, 20 pipets, hypodermic syringes with needles, 1 gal. of seawater, instructions
Sufficient For Class of 30 students
Product Type Sea Urchin Embryology Kit