Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Seed Dispersal Set
SDPSCI_ED

Catalog No. S07537ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
6/Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07537ND 6/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07537ND Supplier Carolina Biological Supply Supplier No. 157970

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • Six types of seed (12 or more of each)
  • Instructions for demonstrating dispersal by mammals: by mechanical propulsion, by water, and by wind

With this set students learn how seeds are dispersed.

Includes 6 types of seeds:

  • Basswood seeds
  • Cockle Burr seeds
  • Maple Wings
  • Thistle pods
  • Sedge seeds
  • Sickle pods and seeds

Specifications

For Use With (Application) Students learn how seeds are dispersed.
Storage Requirements 24°C to 26°C
Quantity 6/Set
Includes 6 types of seed and instructions for demonstrating dispersal by mammals, by mechanical propulsion, by water and by wind (glider, plumed, winged)
Sufficient For Class of 30 students
Product Type Seed Dispersal Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.