Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Termites
SDPSCI_ED

Catalog No. S07666ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
25/Container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07666ND 25/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07666ND Supplier Carolina Biological Supply Supplier No. 143734

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Living and working together!

Experience the highly social and organized interaction of your own termite colony.

  • Low maintenance
  • Great for behavioral studies
  • Excellent for modeling symbiosis
  • With instructions

Specifications

For Use With (Application) Allow students to experience the highly social and organized interaction of own termite colony, low maintenance, great for behavioral studies and excellent for modeling symbiosis.
Storage Requirements 10°C to 15.5°C
Quantity 25/Container
Includes Instructions
Sufficient For Class of 25 students
Product Type Worker Termites
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.