Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Terrestrial Isopods
SDPSCI_ED

Catalog No. S07654ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Pill Bug
Sow Bug
Sow Bug Coupon
Quantity:
25/Container
50/Container
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
S07654ND Pill Bug 50/Container
S07651ND Pill Bug 25/Container
S07274ND Sow Bug 50/Container
S07274CND Sow Bug Coupon 50/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07654ND Supplier Carolina Biological Supply Supplier No. 143082

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

They're crustaceans that live on land - yet breathe through gills!

Sow bugs are fantastic examples of environmental adaptation. Porcellio (sow bugs) will play dead or run away when disturbed. Armadillidium (pill bugs) roll into a protective ball when nudged.

  • Easy to care for in a moist environment
  • Great for teaching evolution, adaptation, behavior, and much more
  • Field collected
  • With instructions

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

For Use With (Application) To teach topics such as evolution, adaptation, behavior and much more, use with AP™ laboratory 11: animal behavior.
Storage Requirements 20°C to 22°C
Quantity 50/Container
Scientific Name Armadillidium
Includes Instructions
Sufficient For Class of 30 students
Product Type Pill Bug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.