Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Tropical Fern Set
SDPSCI_ED

Catalog No. S07571ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
5/Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07571ND 5/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07571ND Supplier Carolina Biological Supply Supplier No. 156842

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • A set of five different tropical fern
  • Interesting forms such as: Boston fern, Staghorn fern, Korean rock fern, Button fern, Lemon button fern, Silver lace fern, Bird's nest fern, etc., may be included

Ferns reproduce via spores and have neither seeds nor flowers. Most ferns have what are called fiddleheads.

Specifications

Storage Requirements 22°C to 25°C
Quantity 5/Set
Includes Set of 5 different tropical ferns, Boston fern, staghorn fern, Korean rock fern, button fern, lemon button fern, silver lace fern, bird's nest fern
Sufficient For Class of 30 students
Product Type Fern Set, Tropical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.