Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Tryptic Soy Agar, Prepared Media Plates 100 x 15mm
SDPSCI_ED

Catalog No. S21801ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
10/Container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S21801ND 10/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21801ND Supplier Carolina Biological Supply Supplier No. 822022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Carefully tested for sterility and to verify their ability to support the characteristic growth of appropriate organisms.

  • Variety of options - bottles, plates, tubes, and dehydrated media
  • Fresh, sterility tested - in stock and ready to ship
  • Free of antibiotics, pesticides, and other chemicals
  • Vast selection of microbiology supplies and cultures available

Note: C of A unavailable for this item

Specifications

Content And Storage 2°C to 8°C
Format Plate
Manufacturing Origin US with Globally Sourced Materials
Media Type Tryptic Soy Agar
Product Type Agar Plate
Sterility Sterile
Target Organism Class Fastidious and Non-fastidious Organisms
Test Type Microbial Cultivation
Quantity 10/Container
Height (Metric) 15 mm
Diameter (Metric) Outside 100 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.