Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Sheep Brain Biosmount™ Preparation
SDPSCI_ED

Catalog No. S22776ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Grade:
6 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S22776ND 6 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S22776ND Supplier Carolina™ Supplier No. 265310

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Dissected to show external and internal parts of sheep brain.

  • Shows two median sagittal sections and two transverse sections
  • All the organs are labeled
  • Size: 5 x 10.5 in.

Specifications

Grade 6 to Undergraduate
Format Set
Product Type Sheep Brain Biosmount
Languages English
Subject Biology - Sheep Brain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.