Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carotegrast methyl | 401905-67-7 | 99.2% | 569.44 | C28H26Cl2N4O5 | 10MM 1ML
SDP

Catalog No. 5000238644
Encompass_Preferred

Carotegrast methyl is an orally active, selective α4 integrin antagonist used in preclinical and translational research to study integrin-mediated inflammation and ulcerative colitis mechanisms. It is supplied as a 10 mM solution in DMSO (1 mL) and as solid powder formats for flexibility in experimental workflows.

  • Orally active α4 integrin antagonist relevant to inflammation research.
  • Available as a 10 mM solution in DMSO and as solid powder formats.
  • High purity suitable for research applications (99.24%).
  • Molecular weight 569.44 g·mol⁻¹ and chemical formula C28H26Cl2N4O5.
  • Useful in models of ulcerative colitis and integrin biology studies.
  • Identified by CAS 401905-67-7 for unambiguous compound reference.

Catalog No. 50-002-38644 Supplier Medchemexpress LLC Supplier No. HY12429010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carotegrast methyl is an orally active, selective α4 integrin antagonist used in preclinical and translational research to study integrin-mediated inflammation and ulcerative colitis mechanisms. It is supplied as a 10 mM solution in DMSO (1 mL) and as solid powder formats for flexibility in experimental workflows.

  • Orally active α4 integrin antagonist relevant to inflammation research.
  • Available as a 10 mM solution in DMSO and as solid powder formats.
  • High purity suitable for research applications (99.24%).
  • Molecular weight 569.44 g·mol⁻¹ and chemical formula C28H26Cl2N4O5.
  • Useful in models of ulcerative colitis and integrin biology studies.
  • Identified by CAS 401905-67-7 for unambiguous compound reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.