Learn More
Medchemexpress LLC HY-124290 5mg , Carotegrast methyl CAS:401905-67-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1242905MG
Encompass_Preferred
HY-124290 5mg Carotegrast methyl CAS: 401905-67-7 Carotegrast methyl (AJM300) is an orally active and selective α4 integrin antagonist. HCA2969, an active metabolite of Carotegrast methyl, is a specific and dual α4β1/α4β7 integrin antagonist. Carotegrast methyl prevents the development of colitis in mice.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9474