Learn More
Medchemexpress LLC β-Carotene-13C10 | 301150-50-5 | 546.80 | 1 MG
SDP
Catalog No. 5000389264
Encompass_Preferred
β-Carotene-13C10 is a 13C-labeled version of β-Carotene, a naturally occurring vitamin A precursor and carotenoid. It modulates reactive oxygen species, demonstrating both antioxidant and anti-inflammatory properties, and can induce apoptosis in breast cancer cells.
  • 13C-labeled compound
  • Acts as a vitamin A precursor
  • Modulates reactive oxygen species
  • Exhibits antioxidant and anti-inflammatory activities
  • Can function as an antioxidant or prooxidant
  • Induces apoptosis in breast cancer cells
  • Useful as a tracer
  • Can be used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-003-89264 Supplier Medchemexpress LLC Supplier No. HYN0411S41MG
β-Carotene-13C10 is a 13C-labeled version of β-Carotene, a naturally occurring vitamin A precursor and carotenoid. It modulates reactive oxygen species, demonstrating both antioxidant and anti-inflammatory properties, and can induce apoptosis in breast cancer cells.
  • 13C-labeled compound
  • Acts as a vitamin A precursor
  • Modulates reactive oxygen species
  • Exhibits antioxidant and anti-inflammatory activities
  • Can function as an antioxidant or prooxidant
  • Induces apoptosis in breast cancer cells
  • Useful as a tracer
  • Can be used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)