Learn More
Medchemexpress LLC β-Carotene | 7235-40-7 | 536.87 | 1 G
SDP
Catalog No. 5000389950
Encompass_Preferred
β-Carotene (Provitamin A) is a naturally-occurring vitamin A precursor and a carotenoid compound. It modulates reactive oxygen species (ROS) and exhibits both antioxidant and anti-inflammatory activities, functioning as either an antioxidant or a prooxidant depending on the biological environment. β-Carotene also demonstrates anticancer activities by inducing apoptosis in breast cancer cells.
  • Naturally-occurring vitamin A precursor.
  • Modulates reactive oxygen species (ROS).
  • Possesses antioxidant and anti-inflammatory activities.
  • Acts as an antioxidant or prooxidant.
  • Induces apoptosis in breast cancer cells.
  • Exhibits anticancer properties.
Catalog No. 50-003-89950 Supplier Medchemexpress LLC Supplier No. HYN04111G
β-Carotene (Provitamin A) is a naturally-occurring vitamin A precursor and a carotenoid compound. It modulates reactive oxygen species (ROS) and exhibits both antioxidant and anti-inflammatory activities, functioning as either an antioxidant or a prooxidant depending on the biological environment. β-Carotene also demonstrates anticancer activities by inducing apoptosis in breast cancer cells.
  • Naturally-occurring vitamin A precursor.
  • Modulates reactive oxygen species (ROS).
  • Possesses antioxidant and anti-inflammatory activities.
  • Acts as an antioxidant or prooxidant.
  • Induces apoptosis in breast cancer cells.
  • Exhibits anticancer properties.