Learn More
Medchemexpress LLC β-Carotene | 7235-40-7 | 536.87 | 100 MG
SDP
Catalog No. 5000389128
Encompass_Preferred
β-Carotene (Provitamin A) is a carotenoid compound and a naturally-occurring precursor to vitamin A. It acts as a modulator of reactive oxygen species (ROS), possessing both antioxidant and anti-inflammatory activities. Its role can vary, acting as either an antioxidant or a prooxidant, depending on its inherent properties and the redox potential of its biological environment.
  • Naturally-occurring precursor to vitamin A
  • Modulator of reactive oxygen species (ROS)
  • Possesses antioxidant and anti-inflammatory activities
  • Can act as an antioxidant or prooxidant depending on environment
  • Induces apoptosis in breast cancer cells
  • Demonstrates anticancer activities
Catalog No. 50-003-89128 Supplier Medchemexpress LLC Supplier No. HYN0411100MG
β-Carotene (Provitamin A) is a carotenoid compound and a naturally-occurring precursor to vitamin A. It acts as a modulator of reactive oxygen species (ROS), possessing both antioxidant and anti-inflammatory activities. Its role can vary, acting as either an antioxidant or a prooxidant, depending on its inherent properties and the redox potential of its biological environment.
  • Naturally-occurring precursor to vitamin A
  • Modulator of reactive oxygen species (ROS)
  • Possesses antioxidant and anti-inflammatory activities
  • Can act as an antioxidant or prooxidant depending on environment
  • Induces apoptosis in breast cancer cells
  • Demonstrates anticancer activities