Learn More
Medchemexpress LLC β-Carotene | 7235-40-7 | 536.87 | 25 G
SDP
Catalog No. 5000389630
Encompass_Preferred
β-Carotene (Provitamin A), a carotenoid compound, is a naturally-occurring vitamin A precursor. β-Carotene is a modulator of reactive oxygen species (ROS), with antioxidant and antiinflammatory activities. It may serve as an antioxidant or as a prooxidant, depending on its intrinsic properties as well as on the redox potential of the biological environment in which it acts. It induces breast cancer cells apoptosis, with anticancer activities.
  • Up-regulates PPAR-γ expression and ROS production in MCF-7 cancer cells.
  • Decreases the survival of MCF-7 cells in a dose-dependent manner.
  • Enhances the expression levels of PPAR-γ mRNA and protein in a time-dependent manner.
  • Down-regulates the COX-2 but up-regulates the p21 mRNA level and protein expression.
  • Increases the percentage of early apoptosis.
  • Induces cytochrome C release.
Catalog No. 50-003-89630 Supplier Medchemexpress LLC Supplier No. 25GHYN0411
β-Carotene (Provitamin A), a carotenoid compound, is a naturally-occurring vitamin A precursor. β-Carotene is a modulator of reactive oxygen species (ROS), with antioxidant and antiinflammatory activities. It may serve as an antioxidant or as a prooxidant, depending on its intrinsic properties as well as on the redox potential of the biological environment in which it acts. It induces breast cancer cells apoptosis, with anticancer activities.
  • Up-regulates PPAR-γ expression and ROS production in MCF-7 cancer cells.
  • Decreases the survival of MCF-7 cells in a dose-dependent manner.
  • Enhances the expression levels of PPAR-γ mRNA and protein in a time-dependent manner.
  • Down-regulates the COX-2 but up-regulates the p21 mRNA level and protein expression.
  • Increases the percentage of early apoptosis.
  • Induces cytochrome C release.