Learn More
Medchemexpress LLC β-Carotene | 7235-40-7 | 536.87 | 50 MG
SDP
Catalog No. 5000390282
Encompass_Preferred
β-Carotene (Standard) is an analytical standard and naturally-occurring vitamin A precursor, primarily intended for research and analytical applications. This carotenoid compound acts as a modulator of reactive oxygen species (ROS), demonstrating both antioxidant and anti-inflammatory activities. It can also induce apoptosis in breast cancer cells, showcasing potential anticancer properties.
Features:
  • Analytical standard for assay reference
  • Modulator of reactive oxygen species (ROS)
  • Exhibits antioxidant and anti-inflammatory activities
  • Can function as an antioxidant or prooxidant
  • Induces apoptosis in breast cancer cells
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-003-90282 Supplier Medchemexpress LLC Supplier No. HYN0411R50MG
β-Carotene (Standard) is an analytical standard and naturally-occurring vitamin A precursor, primarily intended for research and analytical applications. This carotenoid compound acts as a modulator of reactive oxygen species (ROS), demonstrating both antioxidant and anti-inflammatory activities. It can also induce apoptosis in breast cancer cells, showcasing potential anticancer properties.
Features:
  • Analytical standard for assay reference
  • Modulator of reactive oxygen species (ROS)
  • Exhibits antioxidant and anti-inflammatory activities
  • Can function as an antioxidant or prooxidant
  • Induces apoptosis in breast cancer cells
  • Suitable for HPLC, GC, and MS research experiments