Learn More
Medchemexpress LLC β-Carotene (standard) | 7235-40-7 | 536.87 | 100 MG
SDP
Catalog No. 5000389981
Encompass_Preferred
β-Carotene (Standard) is the analytical standard of β-Carotene. This product is intended for research and analytical applications. β-Carotene (Provitamin A), a carotenoid compound, is a naturally-occurring vitamin A precursor. β-Carotene is a modulator of reactive oxygen species (ROS), with antioxidant and antiinflammatory activities. β-Carotene may serve as an antioxidant or as a prooxidant, depending on its intrinsic properties as well as on the redox potential of the biological environment in which it acts. β-Carotene induces breast cancer cells apoptosis, with anticancer activities.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative research experiments in HPLC, GC and MS.
  • It is commonly used in quantitative research experiments in HPLC, GC and MS.
  • It is commonly used in methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-89981 Supplier Medchemexpress LLC Supplier No. HYN0411R100MG
β-Carotene (Standard) is the analytical standard of β-Carotene. This product is intended for research and analytical applications. β-Carotene (Provitamin A), a carotenoid compound, is a naturally-occurring vitamin A precursor. β-Carotene is a modulator of reactive oxygen species (ROS), with antioxidant and antiinflammatory activities. β-Carotene may serve as an antioxidant or as a prooxidant, depending on its intrinsic properties as well as on the redox potential of the biological environment in which it acts. β-Carotene induces breast cancer cells apoptosis, with anticancer activities.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative research experiments in HPLC, GC and MS.
  • It is commonly used in quantitative research experiments in HPLC, GC and MS.
  • It is commonly used in methodological research experiments in HPLC, GC and MS.