Learn More
Medchemexpress LLC Β-carotene (standard) | 7235-40-7 | 536.87 | 25 MG
SDP
Catalog No. 5000389334
Encompass_Preferred
β-Carotene (Standard) is the analytical standard of β-Carotene, intended for research and analytical applications. As a naturally-occurring vitamin A precursor, β-Carotene modulates reactive oxygen species (ROS) with antioxidant and antiinflammatory activities. It may serve as an antioxidant or a prooxidant depending on the biological environment and induces breast cancer cell apoptosis, exhibiting anticancer activities.
  • Analytical standard
  • Intended for research and analytical applications
  • Naturally-occurring vitamin A precursor
  • Modulator of reactive oxygen species (ROS)
  • Antioxidant and antiinflammatory activities
  • May serve as an antioxidant or a prooxidant
  • Induces breast cancer cells apoptosis
  • Anticancer activities
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-003-89334 Supplier Medchemexpress LLC Supplier No. 25MGHYN0411R
β-Carotene (Standard) is the analytical standard of β-Carotene, intended for research and analytical applications. As a naturally-occurring vitamin A precursor, β-Carotene modulates reactive oxygen species (ROS) with antioxidant and antiinflammatory activities. It may serve as an antioxidant or a prooxidant depending on the biological environment and induces breast cancer cell apoptosis, exhibiting anticancer activities.
  • Analytical standard
  • Intended for research and analytical applications
  • Naturally-occurring vitamin A precursor
  • Modulator of reactive oxygen species (ROS)
  • Antioxidant and antiinflammatory activities
  • May serve as an antioxidant or a prooxidant
  • Induces breast cancer cells apoptosis
  • Anticancer activities
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS