Learn More
YMC America, Inc CAROTENOID 3UM 20X2.1MM PK/5
SDP
Catalog No. 50157266
Encompass
CT99S03-02Q1GC: Carotenoid, 3um, 20X2.1mm Cartridge, pk/5
Catalog No. 50-157-266 Supplier YMC America, Inc Supplier No. CT99S0302Q1GC
CT99S03-02Q1GC: Carotenoid, 3um, 20X2.1mm Cartridge, pk/5