Learn More
Cayman Chemical Carotenoid Mixture, 100 mg
SDP
Catalog No. 502966701
Encompass_Preferred
A mixture of carotenoids; contains capsanthin and β-carotene, as well as additional carotenoids and carotenoid esters
Catalog No. 50-296-6701 Supplier Cayman Chemical Supplier No. 18264100 MG
A mixture of carotenoids; contains capsanthin and β-carotene, as well as additional carotenoids and carotenoid esters