Learn More
Cayman Chemical CarotenoId MIxture 10mg
SDP
Catalog No. 502965906
Encompass_Preferred
A mixture of carotenoids; contains capsanthin and β-carotene, as well as additional carotenoids and carotenoid esters
Catalog No. 50-296-5906 Supplier Cayman Chemical Supplier No. 1826410 MG
A mixture of carotenoids; contains capsanthin and β-carotene, as well as additional carotenoids and carotenoid esters