Learn More
Cayman Chemical CarotenoId MIxture 50mg
SDP
Catalog No. 502967142
Encompass_Preferred
A mixture of carotenoids; contains capsanthin and β-carotene, as well as additional carotenoids and carotenoid esters
Catalog No. 50-296-7142 Supplier Cayman Chemical Supplier No. 1826450 MG
A mixture of carotenoids; contains capsanthin and β-carotene, as well as additional carotenoids and carotenoid esters