Learn More
Hach Company Carousel Holder 7 x 1 cm
Catalog No. 501997952 Shop All Hach Company Products
Encompass
The carousel sample changer allows up to seven sequential measurements for mini series or enzymatic methods.
Catalog No. 50-199-7952 Supplier Hach Company Supplier No. LZV902.99.00002
The carousel sample changer allows up to seven sequential measurements for mini series or enzymatic methods.