Learn More
Electron Microscopy Sciences Universal Carousel Carousel Pipette Stand, Assorted/Clear
SDP
Catalog No. 501929486
Encompass_Preferred
Hold and store your pipettes in style in the universal carousel pipette stand. The sleek and modern design of this stand enables it to securely hold virtually every known brand of pipette in the marketplace, making it truly 'universal'. Ideal for labs have multiple brands in use. Hold up to 8 single channel pipettes, 4 multi channel pipettes, or a combination of both pipettes.
Pipettes are held in a vertical position to help prevent contamination. Inserts on the upper holders can be added or removed to accommodate the fit of different pipette widths. One handed removal or replacement of pipettes is easy with the rotating head and stable weighted base. Moving the stand while keeping pipettes in place is simple with a quick grip handle allowing a user to grab and go.
The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants. Made of ABS. US Patent Pending.
Color: Assorted/Clear
Catalog No. 50-192-9486 Supplier Electron Microscopy Sciences Supplier No. 6438703
Hold and store your pipettes in style in the universal carousel pipette stand. The sleek and modern design of this stand enables it to securely hold virtually every known brand of pipette in the marketplace, making it truly 'universal'. Ideal for labs have multiple brands in use. Hold up to 8 single channel pipettes, 4 multi channel pipettes, or a combination of both pipettes.
Pipettes are held in a vertical position to help prevent contamination. Inserts on the upper holders can be added or removed to accommodate the fit of different pipette widths. One handed removal or replacement of pipettes is easy with the rotating head and stable weighted base. Moving the stand while keeping pipettes in place is simple with a quick grip handle allowing a user to grab and go.
The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants. Made of ABS. US Patent Pending.
Color: Assorted/Clear