Learn More
AdipoGen Caroverin
SDP
Catalog No. 502561399
Encompass_Preferred
Chemical. CAS 23465-76-1. Formula C22H27N3O2. MW 365.47. Synthetic. Quinoxaline compound, originally developed as a spasmolytic drug. Acts as an N-type calcium channel blocker, competitive AMPA receptor antagonist, and non-competitive NMDA receptor antagonist. Antioxidant agent through partial prevention of the formation and the highly active scavenging of hydroxyl radicals. Used to treat inner ear diseases such as tinnitus. Shown to inhibit tumor-promoting factors and potentially useful as a chemotherapeutic agent.
Catalog No. 50-256-1399 Supplier AdipoGen Supplier No. CDXC0260M010
Chemical. CAS 23465-76-1. Formula C22H27N3O2. MW 365.47. Synthetic. Quinoxaline compound, originally developed as a spasmolytic drug. Acts as an N-type calcium channel blocker, competitive AMPA receptor antagonist, and non-competitive NMDA receptor antagonist. Antioxidant agent through partial prevention of the formation and the highly active scavenging of hydroxyl radicals. Used to treat inner ear diseases such as tinnitus. Shown to inhibit tumor-promoting factors and potentially useful as a chemotherapeutic agent.