Learn More
AdipoGen Caroverine hydrochloride
SDP
Catalog No. 502561402
Encompass_Preferred
Chemical. CAS 55750-05-5. Formula C22H27N3O2 . HCl . H2O. MW 401.93 . 18.0. Synthetic. Nonselective NMDA and AMPA glutamate receptor antagonist. A class B calcium-channel-blocker, antiglutamatergic agent and shown to act as an antioxidant.
Catalog No. 50-256-1402 Supplier AdipoGen Supplier No. CDXC0258M025
Chemical. CAS 55750-05-5. Formula C22H27N3O2 . HCl . H2O. MW 401.93 . 18.0. Synthetic. Nonselective NMDA and AMPA glutamate receptor antagonist. A class B calcium-channel-blocker, antiglutamatergic agent and shown to act as an antioxidant.