Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Caroverine hydrochloride | 55750-05-5 | 98.9% | 401.93 g/mol | C22H28ClN3O2 | 500 MG
SDP

Catalog No. 5000242110
Encompass_Preferred

Caroverine hydrochloride is the hydrochloride salt of caroverine, a nonselective NMDA and AMPA glutamate receptor antagonist supplied for laboratory research use. It is provided as a dry powder for preparation of solution stocks and for in vitro or in vivo pharmacology studies.

  • CAS number 55750-05-5.
  • Molecular weight 401.93 g/mol.
  • Chemical formula C22H28ClN3O2.
  • Form white to off-white powder.
  • Purity 98.9% (HPLC).
  • Soluble in DMSO and water for stock preparation.
  • Storage store sealed at 4°C, protect from moisture.
  • Research use for NMDA/AMPA receptor pharmacology.

Catalog No. 50-002-42110 Supplier Medchemexpress LLC Supplier No. HY106467B500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Caroverine hydrochloride is the hydrochloride salt of caroverine, a nonselective NMDA and AMPA glutamate receptor antagonist supplied for laboratory research use. It is provided as a dry powder for preparation of solution stocks and for in vitro or in vivo pharmacology studies.

  • CAS number 55750-05-5.
  • Molecular weight 401.93 g/mol.
  • Chemical formula C22H28ClN3O2.
  • Form white to off-white powder.
  • Purity 98.9% (HPLC).
  • Soluble in DMSO and water for stock preparation.
  • Storage store sealed at 4°C, protect from moisture.
  • Research use for NMDA/AMPA receptor pharmacology.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.