Learn More
Medchemexpress LLC Caroverine hydrochloride | 55750-05-5 | MFCD01684929 | 98.9% | 401.93 g/mol | C22H28ClN3O2 | 5 MG
SDP
Catalog No. 5000242108
Encompass_Preferred
Caroverine hydrochloride is a research reagent that acts as a potent, competitive, and reversible antagonist of NMDA and AMPA glutamate receptors. It also exhibits antioxidant and calcium-blocking activity with vasorelaxant effects, and is used in studies such as inner-ear tinnitus research. The compound is supplied as a white to off-white solid with high purity.
  • Antagonist of NMDA and AMPA glutamate receptors.
  • Exhibits antioxidant and calcium-blocking activity.
  • Displays vasorelaxant effects useful in vascular studies.
  • Suitable for research into inner-ear tinnitus mechanisms.
  • High purity for research applications (98.9%).
  • White to off-white solid, stable when stored sealed and protected from moisture.
Catalog No. 50-002-42108 Supplier Medchemexpress LLC Supplier No. HY106467B5MG
Caroverine hydrochloride is a research reagent that acts as a potent, competitive, and reversible antagonist of NMDA and AMPA glutamate receptors. It also exhibits antioxidant and calcium-blocking activity with vasorelaxant effects, and is used in studies such as inner-ear tinnitus research. The compound is supplied as a white to off-white solid with high purity.
  • Antagonist of NMDA and AMPA glutamate receptors.
  • Exhibits antioxidant and calcium-blocking activity.
  • Displays vasorelaxant effects useful in vascular studies.
  • Suitable for research into inner-ear tinnitus mechanisms.
  • High purity for research applications (98.9%).
  • White to off-white solid, stable when stored sealed and protected from moisture.