Learn More
Med Vet International Carpal Arthrodesis Plate, Compression, 2.0/2.0mm, 55mm, J0428Y
Catalog No. 5000582437
Encompass_Preferred
In a carpal arthrodesis the distal screws are often too large on a standard plate for the metacarpal bone. The proximal four holes are larger and the distal four holes are smaller. The middle hole will take either size screw. The plate narrows and has a slight 4° bevel at the distal end.
Catalog No. 50-005-82437 Supplier Med Vet International Supplier No. JOR428Y
In a carpal arthrodesis the distal screws are often too large on a standard plate for the metacarpal bone. The proximal four holes are larger and the distal four holes are smaller. The middle hole will take either size screw. The plate narrows and has a slight 4° bevel at the distal end.