Learn More
Med Vet International Carpal Arthrodesis Plates, Intertarsal Arthrodesis PL, 2/2.7mm, 55mm, J0428Z, Each
Catalog No. 5000582975
Encompass_Preferred
Intertarsal Arthrodesis Compression Plate is slightly wider and the holes are positioned to cover the metatarsal more completely. Middle hole picks up the central tarsal bone and T4. 2.0mm/2.7mm (sheltie size). (not pictured) 55mm.
Catalog No. 50-005-82975 Supplier Med Vet International Supplier No. JOR428Z
Intertarsal Arthrodesis Compression Plate is slightly wider and the holes are positioned to cover the metatarsal more completely. Middle hole picks up the central tarsal bone and T4. 2.0mm/2.7mm (sheltie size). (not pictured) 55mm.