Learn More
Med Vet International Carpal Splint, Small, 10" x 1.25" x 5/8", Each
Catalog No. 5000581393
Encompass_Preferred
These splints are made from a heavier material that is reinforced with longitudinal ribs. A recessed area reduces pressure on a carpal pad. A slight bend in the distal end follows the natural resting position. Splintsare provided with an open pore foam for comfort.
Catalog No. 50-005-81393 Supplier Med Vet International Supplier No. JOR650B
These splints are made from a heavier material that is reinforced with longitudinal ribs. A recessed area reduces pressure on a carpal pad. A slight bend in the distal end follows the natural resting position. Splintsare provided with an open pore foam for comfort.