Learn More
Med Vet International Carpal Splint Toy, 7 1/2"x1"x5/8"D, J0650A
Catalog No. 5000581009
Encompass_Preferred
These splints are made from a heavier material that is reinforced with longitudinal ribs. A recessed area reduces pressure on a carpal pad. A slight bend in the distal end follows the natural resting position. Splintsare provided with an open pore foam for comfort.
Catalog No. 50-005-81009 Supplier Med Vet International Supplier No. JOR650A
These splints are made from a heavier material that is reinforced with longitudinal ribs. A recessed area reduces pressure on a carpal pad. A slight bend in the distal end follows the natural resting position. Splintsare provided with an open pore foam for comfort.