Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carperitide acetate | 1366000-58-9 | 99.8% | 3140.50 | 5 MG
SDP

Catalog No. 5000422326
Encompass_Preferred

Carperitide acetate is a 28-amino acid hormone, normally produced and secreted by the human heart in response to cardiac injury and mechanical stretch. It inhibits endothelin-1 secretion in a dose-dependent way. This product is for research use only and not sold to patients.

  • 28-amino acid hormone
  • Produced and secreted by the human heart
  • Inhibits endothelin-1 secretion
  • Purity of 99.8%
  • Appears as a white to off-white solid
  • Soluble in water (50 mg/mL in H2O)

Catalog No. 50-004-22326 Supplier Medchemexpress LLC Supplier No. HYP1235A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carperitide acetate is a 28-amino acid hormone, normally produced and secreted by the human heart in response to cardiac injury and mechanical stretch. It inhibits endothelin-1 secretion in a dose-dependent way. This product is for research use only and not sold to patients.

  • 28-amino acid hormone
  • Produced and secreted by the human heart
  • Inhibits endothelin-1 secretion
  • Purity of 99.8%
  • Appears as a white to off-white solid
  • Soluble in water (50 mg/mL in H2O)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.