Learn More
MSC Clarke Carpet Extractor, 24 in Cleaning Width, 20 gal Tank Capacity, Battery Powered, Walk Behind
Catalog No. 502838725
Encompass
The Clarke Cleantrack Walk Behind, Battery Carpet Extractor is designed for efficient carpet cleaning in commercial and industrial settings. This robust machine features a 24-inch cleaning width and a 20-gallon solution tank, making it suitable for large areas. Its battery-powered operation provides cordless convenience, while the walk-behind design ensures easy maneuverability.
  • 24 in cleaning width for efficient coverage
  • 20 gal solution tank capacity and 16 gal recovery tank capacity
  • Battery powered for cordless operation
  • 100 psi pump pressure and 93 CFM air flow for thorough cleaning
  • Includes cylindrical brush, electronic float switch kit, hose, and multi-surface brush
Catalog No. 50-283-8725 Supplier MSC Supplier No. 35702034
Add to Cart
Add to Cart
Request a Quote
The Clarke Cleantrack Walk Behind, Battery Carpet Extractor is designed for efficient carpet cleaning in commercial and industrial settings. This robust machine features a 24-inch cleaning width and a 20-gallon solution tank, making it suitable for large areas. Its battery-powered operation provides cordless convenience, while the walk-behind design ensures easy maneuverability.
  • 24 in cleaning width for efficient coverage
  • 20 gal solution tank capacity and 16 gal recovery tank capacity
  • Battery powered for cordless operation
  • 100 psi pump pressure and 93 CFM air flow for thorough cleaning
  • Includes cylindrical brush, electronic float switch kit, hose, and multi-surface brush