Learn More
MSC Clarke CarpetMaster 215, Dual Motor, HEPA Filtration, 1 ea
Catalog No. 502868505
Encompass
The Clarke CarpetMaster 215 is a dual motor upright vacuum designed for efficient cleaning with a 15 in cleaning path. It features HEPA filtration and a disposable bag system, making it suitable for professional use in various environments.
  • Light handle weight for reduced operator fatigue
  • Dual motor design for optimal cleaning power
  • HEPA filtration for effective dust containment
  • Quick-draw detail wand with secure easy-access tools
  • Bag light alerts operator for bag change or clogs
  • Easy access to brush for tool-free change
  • Adjustable height handle and simple controls
  • Equipped with a 50 ft safety yellow power cord for extended reach
  • Onboard storage for tools
Catalog No. 50-286-8505 Supplier MSC Supplier No. 35706100
The Clarke CarpetMaster 215 is a dual motor upright vacuum designed for efficient cleaning with a 15 in cleaning path. It features HEPA filtration and a disposable bag system, making it suitable for professional use in various environments.
  • Light handle weight for reduced operator fatigue
  • Dual motor design for optimal cleaning power
  • HEPA filtration for effective dust containment
  • Quick-draw detail wand with secure easy-access tools
  • Bag light alerts operator for bag change or clogs
  • Easy access to brush for tool-free change
  • Adjustable height handle and simple controls
  • Equipped with a 50 ft safety yellow power cord for extended reach
  • Onboard storage for tools