Learn More
MSC Rochester Midland Corporation Spot/Stain Cleaner, Lemon, 1.0 gal, 1 ea
Catalog No. 502870187
Encompass
This liquid spot/stain cleaner is designed for use on all types of carpeting and upholstery, effectively removing spots, bacteria, and malodors. It features a biodegradable formulation and a refreshing lemon scent.
  • Liquid spot/stain cleaner
  • Biodegradable formulation
  • Lemon scent
  • Removes carpet spots, bacteria, and malodors
  • Suitable for extraction or spin bonnet cleaning
  • Recommended for all types of carpeting and upholstery
  • Primary chemical: Sodium lauryl sulfate
  • Meets CRI pH: 9 standards
  • Supplied in a 1.0 US gallon bottle
Catalog No. 50-287-0187 Supplier MSC Supplier No. 85572931
This liquid spot/stain cleaner is designed for use on all types of carpeting and upholstery, effectively removing spots, bacteria, and malodors. It features a biodegradable formulation and a refreshing lemon scent.
  • Liquid spot/stain cleaner
  • Biodegradable formulation
  • Lemon scent
  • Removes carpet spots, bacteria, and malodors
  • Suitable for extraction or spin bonnet cleaning
  • Recommended for all types of carpeting and upholstery
  • Primary chemical: Sodium lauryl sulfate
  • Meets CRI pH: 9 standards
  • Supplied in a 1.0 US gallon bottle