Learn More
Med Vet International Carprieve Injection, 50mg/mL, 20mL
Catalog No. 502828868
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Carprieve Carprofen Injectable for Dogs contains carprofen, a non-steroidal anti-inflammatory drug (NSAID). Carprieve is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. Each mL of Carprieve Injectable contains 50mg Carprofen. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. Next day air available for an additional charge.
Catalog No. 50-282-8868 Supplier Med Vet International Supplier No. RXCARPRIEVEINJ
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Carprieve Carprofen Injectable for Dogs contains carprofen, a non-steroidal anti-inflammatory drug (NSAID). Carprieve is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. Each mL of Carprieve Injectable contains 50mg Carprofen. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. Next day air available for an additional charge.