Learn More
Apexbio Technology LLC Carprofen 53716-49-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062014
Encompass_Preferred
Carprofen (CAS 53716-49-7) is a non-steroidal anti-inflammatory drug (NSAID) primarily characterized by selective inhibition of cyclooxygenase-2 (COX-2) This selective targeting leads to decreased prostaglandin synthesis mitigating inflammation and pain responses In assays involving canine COX-2 enzyme carprofen exhibits inhibitory activity with an IC50 value of approximately 0 03 mM Due to its targeted COX-2 inhibition carprofen serves as a useful research compound in veterinary and biomedical studies investigating inflammatory pathways and analgesic mechanisms
Catalog No. 50-000-62014 Supplier Apexbio Technology LLC Supplier No. B16905.1
 Restricted Item
Add to Cart
Add to Cart
Carprofen (CAS 53716-49-7) is a non-steroidal anti-inflammatory drug (NSAID) primarily characterized by selective inhibition of cyclooxygenase-2 (COX-2) This selective targeting leads to decreased prostaglandin synthesis mitigating inflammation and pain responses In assays involving canine COX-2 enzyme carprofen exhibits inhibitory activity with an IC50 value of approximately 0 03 mM Due to its targeted COX-2 inhibition carprofen serves as a useful research compound in veterinary and biomedical studies investigating inflammatory pathways and analgesic mechanisms