Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carprofen 98.0+%, TCI America™
SDP

Catalog No. C27011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27011G 1 g
C27015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27011G Supplier TCI America Supplier No. C27011G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53716-49-7
Molecular Formula C15H12ClNO2
Molecular Weight (g/mol) 273.72
MDL Number MFCD00079028
InChI Key PUXBGTOOZJQSKH-UHFFFAOYNA-N
Synonym 6-Chloro-alpha-methyl-9H-carbazole-2-acetic Acid
PubChem CID 2581
ChEBI CHEBI:364453
IUPAC Name 2-(6-chloro-9H-carbazol-2-yl)propanoic acid
SMILES CC(C(O)=O)C1=CC2=C(C=C1)C1=C(N2)C=CC(Cl)=C1

Specifications

Melting Point 200°C
Quantity 1 g
UN Number 2811
Formula Weight 273.72
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carprofen
RTECSNumber FE3180000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.