Learn More
Research Products International Corp Carrageenan, Kappa Type, 250 Grams
SDP
Supplier:  Research Products International Corp C41070250.0
Encompass_Preferred
Kappa carrageenan binds water to form strong, rigid gels in the presence of potassium salts. As the level of potassium is increased, the gel structure becomes tightly aggregated and may result in syneresis (moisture on the gel surface).
Catalog No. 50-488-597