Learn More
Specifications
Specifications
| Chemical Name or Material | Carrez I |
| Color | Yellow |
| CAS | 7732-18-5, 14459-95-1, 7732-18-5 |
| Boiling Point | Approximately 100°C |
| CAS Max % | 3.53%, 96.47% |
| Melting Point | Approximately 0°C |
| CAS Min % | 3.53%, 96.47% |
| Grade | Laboratory |
| Packaging | 500 mL HDPE Bottle |
| Traceability to NIST | NIST SRM# 8040 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
