Learn More
Med Vet International Miltex Amalgam Carrier, Double-Ended, M/L, Tungsten Carbide, Each, 71-62TCA
Supplier:  Med Vet International MIL7162TCA
Encompass_Preferred
Instruments that are used to fill dental cavities with amalgam. Like syringes, they carry the amalgam in the tubular tip which is inserted and pressed in the dental cavity by pressing on the lever which activates the piston inside the tube or the back end of the amalgam. Large. Tungsten carbide. Double-Ended.
Catalog No. 50-264-3509