Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Carrier DNA, 10 mg/mL, Sterile

Catalog No. MP112200205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP112200205 1.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112200205 Supplier MP Biomedicals™ Supplier No. 112200205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Store at 2-8°C.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Biochemicals
Concentration 10 mg/mL
Content And Storage 2°C to 8°C
Quantity 1.25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.