Learn More
CROSSTEX INTERNATIONAL Spore Discs, 3 mm, 100/pk
Catalog No. NC1769746
Encompass_Preferred
Crosstex Inoculated Carrier Spore Discs for monitoring Ethylene Oxide (EO) and Dry Heat processes.
This item is not returnable. View return policy
Catalog No. NC1769746 Supplier CROSSTEX INTERNATIONAL Supplier No. NMDN1806
Crosstex Inoculated Carrier Spore Discs for monitoring Ethylene Oxide (EO) and Dry Heat processes.