Learn More
Bluecatbio Ma Inc CARRIER, STANDARD, 52MM CUTOUT
SDP
Catalog No. NC1532286
Encompass
Additional set of 2 carriers for 14.5 mm plate (height of a standard 96/384-well plate) with 52mm cut-out compatible with HRB grippers.
Catalog No. NC1532286 Supplier Bluecatbio Ma Inc Supplier No. 2CNM14
Additional set of 2 carriers for 14.5 mm plate (height of a standard 96/384-well plate) with 52mm cut-out compatible with HRB grippers.